Learn More
Medchemexpress LLC Interferon alpha-2 (mouse) | 10 UG
SDP
Supplier:  Medchemexpress LLC HYP7544A10UG
Encompass_Preferred
Recombinant mouse interferon-alpha-2 (His-tagged) is a biologically active cytokine produced in E. coli and supplied as a lyophilized powder. It signals via the interferon alpha receptor and is intended for in vitro bioassays and biochemical studies when reconstituted per the datasheet.
  • Lyophilized recombinant protein, suitable for long-term storage.
  • Expressed in E. coli and provided as His-tagged material.
  • High specific activity (ED50 ≈ 0.78 ng/mL) for bioassays.
  • Low endotoxin level (<1 EU/μg) for cell-based applications.
  • Supplied in small vial formats appropriate for assay-scale experiments.
  • Store at -20°C and add carrier protein for extended stability.
Catalog No. 50-002-91327