Learn More
Sigma Aldrich Fine Chemicals Biosciences Interferon-gamma human hIf
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 11040596001
Encompass
IfN-I3 is produced by T-lymphocytes stimulated by antigen or by T-cell mitogens. Under nondenaturating conditions its molecular weight values range from 32 000 to 73 000 indicating that recombinant human IfN-I3 exists as a dimer or higher oligomers.
Catalog No. 50-290-1126