Learn More
Invivogen INTERFERON STIMULATING DNA (IS
SDP
Supplier:  Invivogen TLRLISDN
Encompass
ISD (interferon stimulatory DNA) is a 45-bp non-CpG oligomer from the Listeria monocytogenes genome. When transfected into various cell types, including plasmacytoid and conventional DCs, macrophages and murine embryonic fibroblasts, ISD strongly enhances the expression of IFN-β [1]. This ISD-induced response is mediated by the STING-TBK1-IRF3 signaling axis [1-2]. ISD Naked. Description: CDS Agonist. Unit Size: 200 µg
Catalog No. NC0432595