Learn More
Medchemexpress LLC Interiorin | 119139-55-8 | 482.52 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN119491MG
Encompass_Preferred
Interiorin is a chemical compound isolated from Kadsura heteroclita, known for its moderate anti-HIV activity. This product is intended for research use by qualified scientists in appropriately equipped and authorized facilities. The information provided is based on current knowledge, but the chemical, physical, and toxicological properties have not been completely investigated. The burden of safe use of this material rests entirely with the user.
  • Isolated from Kadsura heteroclita
  • Exhibits moderate anti-HIV activity with an EC50 value of 1.6 μg/mL
  • Intended for research use only
  • Requires handling by suitably qualified and experienced scientists
Catalog No. 50-004-15083