Learn More
Cusabio Technology LLC interleukin 1 alpha
Supplier:  Cusabio Technology LLC CSBE10041MO
Encompass_Preferred
This kit detects IL1A in monkey serum plasma and tissue homogenates offering a broad detection range of 7 8 pg/mL to 500 pg/mL with a sensitivity of 1 95 pg/mL Designed for quantitative analysis it employs a sandwich ELISA methodology ensuring high specificity and accuracy The assay requires an incubation period of 1-5 hours and a sample volume of 50-100 L per test Detection is performed at 450 nm enabling reliable measurement of IL1A concentrations in diverse biological samples Suitable for research applications this kit provides robust performance for monitoring interleukin-1 alpha levels in experimental or diagnostic settings
Catalog No. 50-000-49165