Learn More
Cusabio Technology LLC Bovine Interleukin 1β (IL- 1β) ELISA kit
Supplier:  Cusabio Technology LLC CSBE12986B
Encompass_Preferred
This Bovine IL1B ELISA Kit was designed for the quantitative measurement of Bovine IL1B protein in serum, plasma, tissue homogenates. It is a Sandwich ELISA kit, its detection range is 62.5 pg/mL-4000 pg/mL and the sensitivity is 15.6 pg/mL . One kit contains 96 wells.
Catalog No. NC2952946