Learn More
Cusabio Technology LLC Interleukin 15 IL-15
Supplier:  Cusabio Technology LLC CSBE04603H
Encompass_Preferred
This kit detects IL15 in human serum plasma and tissue homogenates with a sensitivity of 0 78 pg/ml offering a quantitative measurement range of 3 12 pg/ml to 200 pg/ml using a sandwich ELISA methodology The assay requires a sample volume of 50-100 l and can be completed within 1-5 hours Results are measured at an absorbance wavelength of 450 nm providing reliable detection for research or diagnostic applications requiring precise interleukin-15 level analysis The kit is optimized for accuracy and reproducibility across biological matrices making it suitable for studies involving immune response monitoring or cytokine profiling
Catalog No. 50-000-50437