Learn More
Cusabio Technology LLC Human Interleukin 17A (IL-17A/IL-17) ELISA Kit
Supplier:  Cusabio Technology LLC CSBE12819H
Encompass_Preferred
CUSABIO's human IL17A ELISA kit is an in vitro enzyme-linked immunosorbent assay for the quantitative determination of IL17A concentrations in serum, plasma, and tissue homogenates. This assay exclusively recognizes human IL17A. The quantitative sandwich ELISA technique of this kit is based on IL17A antibody-IL17A antigen interactions and an HRP colorimetric detection system to detect the levels of IL17A in samples. The intensity of the color is positively proportional to the amount of bound IL17A in the initial step.
Catalog No. NC3654157