Learn More
Cayman Chemical Interleukin-18 Rabbit Monoclo
SDP
Supplier:  Cayman Chemical 3809250 UL
Encompass_Preferred
A histamine H3 receptor antagonist (Kis 0.13 0.68 and 1.74 nM for the recombinant human rat and mouse receptors respectively) selective for the histamine H3 receptor over a panel of 50 receptors and ion channels at 1 UM prevents imetit-induced decreases in forskolin-stimulated cAMP accumulation in HEK293-Gao cells expressing the human histamine H3 receptor (pA2 9.06) inhibits intake R-(–)-a-methylhistamine-induced increases in water intake and reverses scopolamine-induced memory deficits in the passive avoidance test in rats increases hindlimb grip force in a rat model of monoiodoacetate-induced osteoarthritic pain and increases the paw withdrawal threshold in a rat model of spinal nerve ligation-induced neuropathic pain (ED50s 0.77 and 1.5 mg/kg i.p. respectively)
Catalog No. 50-291-8054