Learn More
Medchemexpress LLC Interleukin-20 receptor beta protein (mouse) | ≥98.0% | 10 UG

Supplier: Medchemexpress LLC HYP7642910UG
Recombinant mouse IL-20 receptor beta protein expressed in HEK293 cells with a C-terminal His tag. Supplied as a purified protein solution (0.36 mg/mL; ~23.8 kDa) with ≥98.0% purity. Store at -80°C for long-term storage; stable at -20°C for up to 3 months after opening. For research use only; shipped on dry ice.
- Recombinant expression in HEK293 cells.
- C-terminal His tag for purification and detection.
- High purity (≥98.0%) suitable for biochemical assays.
- Concentration 0.36 mg/mL; molecular weight ~23.8 kDa.
- Store at -80°C and avoid repeated freeze-thaw; shipped on dry ice.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.