Learn More
Medchemexpress LLC Interleukin-21 (canine) | >95.0% | 20 UG

Supplier: Medchemexpress LLC HYP7481920UG
Recombinant canine interleukin-21 (IL-21) produced in E. coli and supplied as a lyophilized, tag-free protein for immunology and cell biology research. The product is provided in small quantity formats, with documented purity, low endotoxin, and measured biological activity in NK cell assays.
- Lyophilized, tag-free recombinant protein.
- Expressed in E. coli expression system.
- Purity greater than 95.0% by reducing SDS-PAGE.
- Endotoxin less than 1 EU/μg (LAL method).
- Sequence corresponds to mature protein residues H18-S146.
- Predicted molecular mass ~15 kDa.
- Biological activity with ED50 approximately 7.8 ng/mL in NK-92 assays.
- Recommend reconstitution to at least 100 μg/mL in ddH2O.
- Store at -20 °C; stable for 2 years from receipt.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.