Learn More
Cusabio Technology LLC Rat Interleukin 6,IL-6 ELISA KIT
Supplier:  Cusabio Technology LLC CSBE04640R
Encompass_Preferred
This Rat IL6 ELISA Kit was designed for the quantitative measurement of Rat IL6 protein in serum, plasma, tissue homogenates. It is a Sandwich ELISA kit, its detection range is 0.312 pg/mL-20 pg/mL and the sensitivity is 0.078 pg/mL. One kit contains 96 wells.
Catalog No. NC2952868