Learn More
Cusabio Technology LLC Human Interleukin 6,IL-6 ELISA KIT
Supplier:  Cusabio Technology LLC CSBE04638H
Encompass_Preferred
The human IL-6 ELISA kit is suitable for the quantitative determination of human IL-6 in a wide range of sample types, including serum, plasma, cell culture supernates, tissue homogenates, and urine. This assay employs the bi-antibody sandwich technique and enzyme-substrate chromogenic reaction to quantify antigen levels in the sample. The amount of synthesized colored products are positively related to the analyte of interest in the sample.This human IL-6 ELISA kit has been described in up to 143 publications. One kit contains 96 wells.
Catalog No. NC2952796