Learn More
Creative Biomart Interleukin-8 (IL-8) Protein
Supplier:  Creative Biomart THP0255
Encompass_Preferred
Interleukin 8 (IL-8 or CXCL8) is a member of the CXC cytokine family and is produced by macrophages, epithelial, smooth muscle, and endothelial cells. IL-8 binds the G protein-coupled serpentine receptors CXCR1 and CXCR2. IL-8 recruits innate immune cells, induces phagocytosis, and stimulate angiogenesis. Recombinant human Interleukin 8 is a non-glycosylated protein monomer, containing 77 amino acids and having a molecular mass of 8.9 kDa.
Catalog No. 50-208-4306