Learn More
Apexbio Technology LLC Interleukin II (60-70) 1mg
SDP
Supplier:  Apexbio Technology LLC A10301
Encompass_Preferred
Interleukin II 60-70 is a cytokine-derived peptide fragment It is designed to modulate immune system activity by influencing lymphocyte function and T-cell differentiation pathways Interleukin II 60-70 exerts its biological activity primarily through the regulation of leukocyte particularly lymphocyte proliferation growth and differentiation of T cells into effector subsets as well as supporting regulatory T cell (Treg) development necessary for immune tolerance Based on these pharmacological properties Interleukin II 60-70 holds research potential in studies of lymphocyte activation mechanisms T-cell differentiation and immune tolerance
Catalog No. 50-000-92966