Learn More
Akron Biotechnology LLC Recombinant Human Interleukin-12 Lyophilized, GMP Grade, 200 μg vial, AR1051-0200
Supplier:  Akron Biotechnology LLC AR10510200
Encompass
Akron's Recombinant Human Interleukin-12 (rHu IL-12) products are manufactured following all relevant cGMP guidelines for ancillary materials Product Features:Active Substance•Carrier protein-free formulation•Accurate mass and activity control•High Purity via RP-HPLC (≥ 95%)Manufacturing•Multi-step downstream purification strategy excluding affinity tags•Sterile microfiltration and aseptic filling/lyophilization•Animal free expression system with yeast hostStability• Under long-term stability•Store at -20 °C•Transport on dry iceFor Use StatementFor research use or further manufacturing use in ex vivo cell therapy applications. This product is not intended for direct in vivo use or for direct clinical use as a drug, therapeutic, biologic, or medical device.
Catalog No. NC2941870
Only null left
Add to Cart
Request a Quote