Learn More
Medchemexpress LLC Intermediate capsid 50ug
SDP
Supplier:  Medchemexpress LLC HYP7230250UG
Encompass_Preferred
The intermediate capsid protein VP6 self-assembles into an icosahedral capsid with T 13 symmetry forming the middle layer of the virus particle It contains 230 VP6 trimers that protect dsRNA and prevent host cell antiviral responses Intermediate capsid protein VP6 Rotavirus A (sf9 His myc ) is the recombinant Virus-derived Intermediate capsid protein VP6 expressed by Sf9 insect cells with N-His C-Myc labeled tag
Catalog No. 50-001-78371