Learn More
Norell Inc 5mm, 500MHz 7" VALVED THIN WALL NMR TUBE FOR INTERMEDIATE PRESSURE 1 Tube per pack
SDP
Supplier:  Norell Inc S5500IPV7
Encompass_Preferred
These pressure tubes facilitate experiments requiring conditions such as pressurized inert atmosphere blanketing, addition of reactive gaseous reagents under pressure, containment of low boiling point solvents or samples at elevated temperatures, and so on.We recommend that the maximum operating pressure should be limited to 600 kPa (6 bar, 87 psi) when using a thin wall pressure tube, up to 1200 kPa (12 bar, 175 psi) if using a heavy wall pressure tube. (Please see accompanying table for complete details).
Catalog No. NC2832782