Learn More
Thermco Products Inc Min-Max Memory Digital Thermometer with NIST Traceable Certificate, Dual Temperature Display, Internial Unit Sensor Range 0/50C (32/122F), External Probe Range -50/70C (-58/158F) , Accuracy: +/- 1.0C (1.8F)
Supplier:  Thermco Products Inc ACC800DIG
Encompass_Preferred
This product was recently added by customer request, and is available for your convenience. We strive to provide our customers with a one-stop shop for the entire scientific supplies category. More relevant content may be added as customer demand increases.
Catalog No. NC0805492