Learn More
Maine Molecular Quality Controls Inc INTROL CF PANEL 3X1ML
SDP
Supplier:  Maine Molecular Quality Controls Inc G106AC1
Encompass
INTROL CF Panel I Control v.02 is intended for in vitro diagnostic use as a quality control to monitor analytical performance of the extraction, amplification and detection steps of diagnostic assays used in the detection of the Cystic Fibrosis Transmembrane Conductance Regulator (CFTR) gene mutations and variants. This product is intended to be extracted and analyzed routinely with each CFTR assay run. The INTROL CF Panel I Control v.02 is designed to monitor the detection of 38 CFTR mutations associated with cystic fibrosis, including the 23 mutations recommended for testing by American College of Medical Genetics (ACMG) and American College of Obstetricians and Gynecologists (ACOG). The INTROL CF Panel I Control also monitors 3 polymorphisms (I506V, I507V, F508C) and the 5/7/9T variants. G106ac-1 Kit Contains: 3 bottles x 1mL 1 each G106a-1, G106b-1, and G106c-1.
Catalog No. NC1935165