Learn More
Medchemexpress LLC Inulicin (1-O-Acetylbritannilactone) | 33627-41-7 | 308.37 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN08961MG
Encompass_Preferred
Inulicin (1-O-Acetylbritannilactone) is an active compound that inhibits VEGF-mediated activation of Src and FAK, and also inhibits LPS-induced PGE2 production and COX-2 expression, as well as NF-κB activation and translocation. This compound is suitable for research applications and has shown diverse biological activities.
  • Inhibits VEGF-mediated activation of Src and FAK.
  • Inhibits LPS-induced PGE2 production and COX-2 expression.
  • Inhibits NF-κB activation and translocation.
  • Inhibits angiogenesis and lung cancer cell growth.
  • Exhibits cytotoxicity against human HCT116, HeLa, and SGC7901 cells.
  • Suppresses A549 xenograft growth in nude mice.
Catalog No. 50-004-16842