Learn More
Medchemexpress LLC Inupadenant hydrochloride | 2411004-22-1 | 99.6% | C25H27ClF2N8O4S2 | 10MG

Supplier: Medchemexpress LLC HY137442A10MG
Inupadenant hydrochloride is an orally active, highly selective adenosine A2A receptor antagonist provided as the hydrochloride salt for research use. It is reported to be non-brain-penetrant and is used in preclinical pharmacology studies to investigate A2A receptor modulation.
- High purity: 99.6%.
- Selective A2A receptor antagonist for receptor pharmacology studies.
- Non-brain-penetrant, suitable for peripheral pharmacology models.
- Physical form: solid, off-white to light yellow.
- Recommended storage: store under inert atmosphere at 4°C; in solution, -80°C (6 months) or -20°C (1 month).
- Intended for research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.