Learn More
Sigma Aldrich Fine Chemicals Biosciences Invertase Activity Assay K
Supplier:  Sigma Aldrich Fine Chemicals Biosciences MAK1181KT
Encompass
Invertase (beta-fructofuranosidase) is an enzyme that catalyzes the hydrolysis of sucrose to fructose and glucose. Invertase is produced by certain plants microorganisms and honey bees.
Catalog No. 50-290-4436