Learn More
Bio X Cell InVivoMAb Anti-Mouse/Human Phosphorylated PD-1 (CD279), Clone 407.6G12 - 1 mg
Supplier:  Bio X Cell BE0387R001MG
Encompass_Preferred
Monoclonal antibody that specifically recognizes PD-1 only when tyrosine 248 (Y248) is phosphorylated. Suitable for detecting phospho-PD-1 in mouse and human samples and for evaluating PD-1 pathway blockade.
  • Specifically recognizes PD-1 phosphorylated at tyrosine 248 (Y248).
  • Reactive with mouse and human PD-1 when phosphorylated.
  • Formulated in PBS, pH 7.0, without stabilizers or preservatives.
  • Concentration is lot specific; generally 4-11 mg/ml.
  • Intended for applications assessing PD-1 phosphorylation and pathway blockade.
Catalog No. NC2236462