Learn More
BIOHIPPO InVivoMAb Anti-F8/Coagulation factor VIII Antibody (KM41), 100 µg
SDP
Supplier:  BIOHIPPO BHA21409798
Encompass_Preferred
F8/Coagulation factor VIII Recombinant Antibody (clone KM41) is a recombinant Mouse IgG2a, kappa antibody specific for F8/Coagulation factor VIII. It reacts with Human. Suitable for ELISA, Neutralization. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.
Catalog No. 50-008-61001