Learn More
BIOHIPPO InVivoMAb Anti-Hepatitis E virus/HEV pORF2 P domain Neutralizing Antibody (Iv0278), 100 µg
SDP
Supplier:  BIOHIPPO BHA21409502
Encompass_Preferred
Hepatitis E virus/HEV pORF2 P domain Neutralizing Recombinant Antibody (clone Iv0278) is a recombinant Human IgG1, kappa antibody specific for Hepatitis E virus/HEV pORF2 P domain Neutralizing. It reacts with Hepatitis E virus/HEV. Suitable for Blocking, Neutralization, ELISA, SPR. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.
Catalog No. 50-008-61284