Learn More
BIOHIPPO InVivoMAb Anti-HRSV/hMPV Fusion glycoprotein Neutralizing Antibody (M2D2), 100 µg
SDP
Supplier:  BIOHIPPO BHA21409531
Encompass_Preferred
HRSV/hMPV Fusion glycoprotein Neutralizing Recombinant Antibody (clone M2D2) is a recombinant Human IgG1, kappa antibody specific for HRSV/hMPV Fusion glycoprotein Neutralizing. It reacts with Human respiratory syncytial virus. Suitable for ELISA, Neutralization. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.
Catalog No. 50-008-60918