Learn More
BIOHIPPO InVivoMAb Anti-Human ARG1/Arginase-1 Antibody (Iv0120), 100 µg
SDP
Supplier:  BIOHIPPO BHA21409876
Encompass_Preferred
ARG1/Arginase-1 Recombinant Antibody (clone Iv0120) is a recombinant Mouse IgG2a, kappa antibody specific for ARG1/Arginase-1. It reacts with Human. Suitable for ELISA, FCM, Neutralization. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.
Catalog No. 50-008-60021