Learn More
BIOHIPPO InVivoMAb Anti-Human TNFSF9/4-1BBL Antibody (Iv0139), 100 µg
SDP
Supplier:  BIOHIPPO BHA21409850
Encompass_Preferred
TNFSF9/4-1BBL Recombinant Antibody (clone Iv0139) is a recombinant Mouse IgG2a, kappa antibody specific for TNFSF9/4-1BBL. It reacts with Human. Suitable for ELISA, FCM, Neutralization. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.
Catalog No. 50-008-60065