Learn More
Bio X Cell InVivoMAb Anti-Mouse DR5 (CD262) Monoclonal Antibody, Clone MD5-1, 50 mg
Supplier: Bio X Cell BE0161R050MG
InVivoMAb anti-mouse DR5 (CD262) is a purified monoclonal antibody (clone MD5-1) intended for in vivo and in vitro induction of TRAIL-mediated apoptosis in mouse models. It is supplied in an in vivo-grade purified format suitable for murine functional studies.
- Purified monoclonal antibody (clone MD5-1).
- Target: mouse death receptor 5 (DR5, CD262).
- Species reactivity: mouse.
- Induces TRAIL-mediated apoptosis in vitro and in vivo.
- Available in 50 mg format for in vivo dosing.
- Produced in an animal-free facility and purified from cell culture supernatant.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.