Learn More
Bio X Cell InVivoMAb Anti-Mouse IL-1 R (CD121a), 100 MG
Supplier: Bio X Cell BE0256R100MG
The InVivoMAb anti-mouse IL-1 R (CD121a) antibody is a monoclonal antibody that reacts with mouse IL-1 receptor type 1. This 80 kDa transmembrane glycoprotein is expressed on various cell types and mediates IL-1 biological responses upon ligand binding.
- Reacts with mouse IL-1 receptor type 1 (CD121a)
- Monoclonal antibody targeting IL-1 receptor
- Transmembrane glycoprotein, 80 kDa
- Binds IL-1α and IL-1β
- Mediates all known IL-1 biological responses
- Purified from cell culture supernatant in an animal-free facility
- Purity ≥95% (SDS-PAGE)
- Endotoxin ≤1EU/mg (LAL assay)
- Sterility: 0.2 μm filtration
- Storage at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.