Learn More
Bio X Cell InVivoMAb anti-mouse Ly6G, Clone 1A8, 1 MG
Supplier: Bio X Cell BE0075R001MG
The 1A8 monoclonal antibody specifically reacts with mouse Ly6G, a 21–25 kDa GPI-anchored cell surface protein involved in cell signaling and adhesion. This antibody is a marker for mature granulocytes and peripheral neutrophils, and specifically targets Ly6G without reported cross-reactivity with Ly6C.
- Clone: 1A8
- Isotype: Rat IgG2a, κ
- Reactivity: Mouse
- Formulation: PBS, pH 7.0; no stabilizers or preservatives
- Endotoxin: ≤1 EU/mg (≤0.001 EU/μg)
- Purity: ≥95% (SDS-PAGE)
- Sterility: 0.2 μm filtration
- Purification method: Protein G
- Antibody molecular weight: 150 kDa
- Applications include in vivo neutrophil depletion, immunofluorescence, and flow cytometry
- Storage: Store at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.