Learn More
Bio X Cell InVivoMAb Anti-Rat Kappa Immunoglobulin Light Chain, 25 mg (Clone MAR 18.5)
Supplier: Bio X Cell BE0122R025MG
InVivoMAb anti-rat Kappa Immunoglobulin Light Chain is an unconjugated mouse monoclonal antibody (clone MAR 18.5) that binds the kappa light chain of rat immunoglobulins. It is supplied as a purified, sterile formulation intended for in vivo use and immunological experiments, provided as a 25 mg vial.
- Unconjugated mouse monoclonal antibody (clone MAR 18.5).
- Targets rat immunoglobulin kappa light chain.
- Supplied as a 25 mg vial in PBS, pH 7.0.
- Purified by Protein G; purity ≥95%.
- Endotoxin ≤1 EU/mg.
- Sterile filtered through 0.2 μm.
- Store at 4°C; do not freeze.
- Suitable for in vivo B cell depletion and immunological assays.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.