Learn More
Bio X Cell InVivoPlus Anti-Mouse CTLA-4 (CD152, Clone 9D9), 50 mg
Supplier: Bio X Cell BP016450MG
InVivoPlus Anti-Mouse CTLA-4 (CD152, clone 9D9) is an ultrapure monoclonal antibody formulated for in vivo and in vitro immune modulation studies. It binds mouse CTLA-4 (CD152) and has been used for CTLA-4 neutralization and intra-tumoral regulatory T cell depletion.
- Clone: 9D9
- Isotype: Mouse IgG2b
- Species reactivity: Mouse
- Applications: In vivo neutralization, intra-tumoral regulatory T cell depletion, Western blot
- Format: Unconjugated; PBS, pH 7.0; no stabilizers or preservatives
- Purity: ≥95%
- Endotoxin: ≤0.5 EU/mg (≤0.0005 EU/μg)
- Available sizes: 5 mg, 25 mg, 50 mg, 100 mg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.