Learn More
Bio X Cell Anti-Human IgE, Omalizumab Biosimilar, Unconjugated, 100 MG
Supplier: Bio X Cell SIM0016R100MG
This non-therapeutic biosimilar antibody utilizes the same variable regions as the therapeutic antibody Omalizumab, making it suitable for research applications. It reacts with free human IgE in blood and interstitial fluid, and with membrane-bound IgE on IgE-expressing B cells. It does not bind to IgE complexed with FcεRI and does not induce histamine release.
- Target human IgE
- Human species reactivity
- Unconjugated
- Human IgG1, κ isotype
- Immunogen human IgE
- Purity ≥95% by SDS-PAGE
- Endotoxin ≤0.5EU/mg by LAL assay
- Molecular weight 150 kDa
- Formulation PBS, pH 7.0, no stabilizers or preservatives
- Store at 4°C, do not freeze
- Applications: IgE neutralization, flow cytometry, ELISA
- Purification by protein A
- Sterility by 0.2 μm filtration
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.