Learn More
Bio X Cell Anti-human IgE, Omalizumab, IgG1 kappa, 5 MG
Supplier: Bio X Cell SIM0016R005MG
This non-therapeutic biosimilar antibody uses Omalizumab's variable regions for research applications. It reacts with free human IgE in blood and interstitial fluid, and membrane-bound IgE on B cells. It does not bind IgE already bound by FcεRI on mast cells, basophils, and dendritic cells, nor does it provoke histamine release. Omalizumab binds an epitope near the FcεRI binding site.
- Isotype: Human IgG1, κ
- Immunogen: Human IgE
- Reported applications: IgE neutralization, Flow cytometry, ELISA
- Formulation: PBS, pH 7.0; Contains no stabilizers or preservatives
- Endotoxin: ≤0.5EU/mg (≤0.0005EU/μg)
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Purification: Protein A
- Molecular Weight: 150 kDa
- Storage: Store antibody solution at stock concentration at 4°C. Do not freeze.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.