Learn More
Bio X Cell InVivoSIM VISTA Antibody, Onvatilimab Biosimilar, 5 MG
Supplier: Bio X Cell SIM0057R005MG
This biosimilar antibody, derived from Onvatilimab, is a humanized IgG1 monoclonal antibody for research applications, reacting with V-type immunoglobulin domain-containing suppressor of T-cell activation (VISTA), also known as VSIR or B7-H5.
- Isotype: Human IgG1, κ
- Reactivity: Human, Monkey
- Immunogen: Human VISTA
- Applications: in vitro and in vivo VISTA signaling blockade, functional assays, flow cytometry, ELISA
- Formulation: PBS, pH 7.0 (no stabilizers or preservatives)
- Endotoxin: ≤0.5 EU/mg
- Purity: ≥95% (by SDS-PAGE)
- Sterility: 0.2 μm filtration
- Production: Animal-free facility
- Purification: Protein A
- Molecular Weight: 150 kDa
- Storage: Store at stock concentration at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.