Learn More
Bio X Cell InVivoMAb anti-mouse MAdCAM-1 (MECA-367), Rat IgG2a , purified, 5 MG, -15MG
Supplier: Bio X Cell BE0035R005MG
Purified rat monoclonal antibody (clone MECA-367) that recognizes and neutralizes mouse MAdCAM-1 (mucosal addressin cell adhesion molecule-1). Supplied as an unconjugated, low-endotoxin preparation suitable for in vivo blocking and immunofluorescence applications. Supplied in a 5 mg vial; store at 4C and do not freeze.
- Clone: MECA-367.
- Host: rat.
- Isotype: IgG2a, .
- Format: purified, unconjugated antibody.
- Target: mouse MAdCAM-1 (functional blocking).
- Purity: 95% (SDS-PAGE).
- Buffer: PBS, pH 7.0.
- Endotoxin: 1 EU/mg (0.001 EU/g).
- Sterility: 0.2 m filtered.
- Storage: store at 4C at stock concentration; do not freeze.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.