Learn More
Medchemexpress LLC INX-P | 2734878-02-3 | 99.6% | 866.79 g/mol | C43H52BrN3O11 | 5 MG

Supplier: Medchemexpress LLC HY1601825MG
INX-P is a glucocorticosteroid-target antibody-drug conjugate (ADC) supplied for research use only. It is characterized by high purity and is intended for preclinical and laboratory studies.
- Glucocorticosteroid-target ADC for research use only
- Purity 99.63%
- CAS number 2734878-02-3
- Molecular weight 866.79 g/mol
- Chemical formula C43H52BrN3O11
- Available in small quantities for experimental applications
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.