Learn More
Medchemexpress LLC Iobenguane (sulfate) | 87862-25-7 | 99.8% | 10 MM 1 ML
SDP
Supplier:  Medchemexpress LLC HY129040A10MM/1ML
Encompass_Preferred
m-Iodobenzylguanidine sulfate (MIBG sulfate) is an analogue of the neurotransmitter norepinephrine with antitumor activity. Radioiodinated m-Iodobenzylguanidine sulfate is clinically used as a tumor-targeted radiopharmaceutical in the diagnosis and treatment of adrenergic tumors. It is also a high-affinity substrate for cholera toxin that interferes with cellular mono(ADP-ribosylation).
  • Analogue of neurotransmitter norepinephrine
  • Possesses antitumor activity
  • Used as a tumor-targeted radiopharmaceutical in diagnosis and treatment of adrenergic tumors
  • High-affinity substrate for cholera toxin
  • Interferes with cellular mono(ADP-ribosylation)
Catalog No. 50-003-38845