Learn More
Revvity Health Sciences Inc Iodine-125 Radionuclide, 10mCi, Specific Activity: ~17Ci/mg, 0.1M NaOH (pH 12-14), Concentration: >350mCi/mL
Supplier:  Revvity Health Sciences Inc NEZ033H010MC
Encompass
Iodine-125 Radionuclide, 10mCi, Specific Activity: ~17Ci/mg, 0.1M NaOH (pH 12-14), Concentration: >350mCi/mL
Catalog No. 50-280-8892