Learn More
Revvity Health Sciences Inc Iodine-125 Radionuclide, 2mCi, Specific Activity: ~17Ci/mg, 10-5M NaOH (pH 8-11)
Supplier:  Revvity Health Sciences Inc NEZ033A002MC
Encompass
Iodine-125 Radionuclide, 2mCi, Specific Activity: ~17Ci/mg, 10-5M NaOH (pH 8-11)
Catalog No. 50-280-9402