Learn More
Medchemexpress LLC Iodipamide (Adipiodone) | 606-17-7 | 100.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB129210MM/1ML
Encompass_Preferred
Iodipamide (Adipiodone) is an ionic iodinated contrast agent primarily used for diagnostic imaging to visualize bile ducts and the gallbladder during X-ray examinations. After intravenous administration, it is transported to the liver and rapidly excreted into bile, enabling targeted contrast examinations.
  • Ionic iodinated contrast agent
  • Used for diagnostic imaging
  • Visualizes bile ducts and gallbladder
  • Enables targeted contrast examinations
  • Intended for research use only
Catalog No. 50-004-13210