Learn More
Medchemexpress LLC Iodixanol | 92339-11-2 | 100.0% | 250 MG

Supplier: Medchemexpress LLC 250MGHYB1426
Iodixanol is an iodine-containing non-ionic radiocontrast agent, offering a high purity of 99.97%. It is supplied as a solid, with specific storage recommendations to ensure stability and efficacy.
- Non-ionic radiocontrast agent.
- Purity of 99.97%.
- Supplied as a solid.
- Features specific storage conditions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.