Learn More
Medchemexpress LLC Iodoacetamide-PEG5-azide | 98.6% | 474.29 g/mol | C14H27IN4O6 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1346875MG
Encompass_Preferred
Iodoacetamide-PEG5-azide is a PEG-based linker and click-chemistry reagent bearing an iodoacetamide and an azide functional group. It is intended for bioconjugation and PROTAC synthesis workflows, enabling selective cysteine modification via the iodoacetamide and subsequent conjugation through azide-alkyne cycloaddition.
  • Contains iodoacetamide and azide functional groups.
  • PEG5 spacer provides flexible linker length.
  • Compatible with copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Also compatible with strain-promoted alkyne-azide cycloaddition (SPAAC).
  • High purity (98.6%) suitable for research applications.
Catalog No. 50-002-21871