Learn More
Medchemexpress LLC Iodoacetyl-LC-biotin | 93285-75-7 | MFCD00054987 | 99.2% | 510.43 g/mol | C18H31IN4O3S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1380655MG
Encompass_Preferred
Iodoacetyl-LC-biotin is a thiol-reactive, biotinylated electrophile reagent used for site-specific labeling and enrichment of cysteine-containing proteins. It reacts with sulfhydryl groups to form stable thioether bonds, enabling affinity capture and detection in proteomics and biochemical assays.
  • Thiol-specific labeling of cysteine residues for targeted modification.
  • Forms stable thioether bonds for irreversible protein labeling.
  • Enables affinity enrichment and sensitive detection of modified proteins.
  • Compatible with proteomics workflows and subcellular proteome analysis.
  • Supplied as a solid with high purity for research applications.
Catalog No. 50-002-22879