Learn More
Medchemexpress LLC Iodobenzene | 591-50-4 | MFCD00001029 | 100.0% | 204.01 g/mol | C6H5I | 1mL
SDP
Supplier:  Medchemexpress LLC HYY074710MM/1ML
Encompass_Preferred
Iodobenzene can undergo Ullmann coupling reaction on copper catalyst to generate biphenyl Iodobenzene can undergo Suzuki reaction with phenylboronic acid to generate biphenyl[1][2]
Catalog No. 50-002-68590