Learn More
Medchemexpress LLC Iodopropynyl butylcarbamate | 55406-53-6 | 99.5% | 281.09 | 25 G
Supplier: Medchemexpress LLC 25GHYW009335
Iodopropynyl butylcarbamate (IPBC) is a potent fungicide and preservative. It is utilized across various industries including paints, coatings, wood preservatives, personal care, and cosmetics. This substance is designated for laboratory chemical use and the manufacture of other substances, intended strictly for research purposes by qualified personnel.
- Potent fungicide and preservative
- Used in paints, coatings, wood preservatives, personal care, and cosmetics industries
- Identified for laboratory chemical use
- Intended for research use only