Learn More
Selleck Chemical LLC Iohexol S4531-100mg
SDP
Supplier:  Selleck Chemical LLC S4531100MG
Encompass_Preferred
Iohexol (Omnipaque) is a radiographic contrast agent that can be used as an exogenous filtration marker for accurate glomerular filtration rate (GFR) estimates
Catalog No. 50-311-1385