Learn More
Apexbio Technology LLC Iohexol 66108-95-0 500mg
SDP
Supplier:  Apexbio Technology LLC B3538500
Encompass_Preferred
Iohexol (CAS 66108-95-0) is a non-ionic hydrophilic iodine-containing contrast agent employed primarily in diagnostic radiology Due to its capability to absorb X-rays effectively iohexol enhances visualization of bodily structures through radiographic imaging techniques such as myelography nephroangiography arthrography and arteriography Its mechanism of action relies upon high radiodensity induced by iodine atoms resulting in clear contrast differentiation between tissues The compound s minimal systemic toxicity arises from its low chemotoxicity and reduced osmolality making it suitable for biomedical research involving imaging-based analysis of anatomical structures and vascular assessments
Catalog No. 50-000-64248